missing translation for 'onlineSavingsMsg'
Learn More

p-Aminobenzoic Acid, 99+%, Free Acid, MP Biomedicals™

Product Code. 11458190 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458190 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11458190 Supplier MP Biomedicals Supplier No. 0210256990
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 150-13-0
Color White
Density 1.374g/mL
Assay Percent Range 99+%
Molecular Formula C7H7NO2
MDL Number MFCD00007894
Quantity 500 g
Synonym p-aminobenzoic acid, paba, para-aminobenzoic acid, vitamin bx, sunbrella, p-carboxyaniline, 4-carboxyaniline, hachemina, paraminol, benzoic acid, 4-amino
InChI Key ALYNCZNDIQEVRV-UHFFFAOYSA-N
SMILES NC1=CC=C(C=C1)C(O)=O
IUPAC Name 4-aminobenzoic acid
Molecular Weight (g/mol) 137.14
PubChem CID 978
ChEBI CHEBI:30753
Formula Weight 137.1
Physical Form Solid
Chemical Name or Material p-Aminobenzoic Acid, Free Acid
Show More Show Less
compliance-icons
Product Identifier
  • p-Aminobenzoic Acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.