missing translation for 'onlineSavingsMsg'
Learn More

p-Acetotoluidide, 99%

Product Code. p-3735225 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10618914

Brand: Thermo Scientific Acros 400161000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-89-9 | C9H11NO | 149.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-89-9
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00008677
InChI Key YICAMJWHIUMFDI-UHFFFAOYSA-N
Synonym 4'-methylacetanilide, p-acetotoluidide, n-4-methylphenyl acetamide, 4-methylacetanilide, n-acetyl-p-toluidine, p-acetotoluide, acetyl-p-toluidine, p-acetotoluidine, acetamide, n-4-methylphenyl, 4-acetotoluide
PubChem CID 7684
IUPAC Name N-(4-methylphenyl)acetamide
SMILES CC1=CC=C(C=C1)NC(=O)C

Specifications

Melting Point 148.0°C to 151.0°C
Color Beige to Brown
Density 1.2100g/mL
Boiling Point 307.0°C
Flash Point 168°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4NHCOCH3
Quantity 100 g
Merck Index 15, 72
Specific Gravity 1.21
Solubility Information Solubility in water: 1g/l(25°C). Other solubilities: soluble in glacial acetic acid,soluble in alcohol,ether,ethyl acetate
Formula Weight 149.19
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material p-Acetotoluidide
Show More Show Less
compliance-icons
Product Identifier
  • p-Acetotoluidide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.