missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ P-5 - Pipette Shelf for UV Cabinet

Product Code. 17593926
Change view
Click to view available options
For Use With (Equipment):
UVC/T-M-AR, UVT-B-AR or UVT-S-AR
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17593926 UVC/T-M-AR, UVT-B-AR or UVT-S-AR Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17593926 Supplier Grant Instruments™ Supplier No. P5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

For Use With (Equipment) UVC/T-M-AR, UVT-B-AR or UVT-S-AR
Height (Metric) 50 mm
Dimensions 140 x 230 x 50 mm
No. of Shelves 1
Depth (Metric) 140 mm
Description P-5, Shelf for Pipettes
Capacity (Metric) Max. Load 5
Color Transparent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.