missing translation for 'onlineSavingsMsg'
Learn More

P-[2-Hydroxy-1-(phenylmethoxy)propyl]phosphonic Acid Dimethyl Ester, TRC

Product Code. 30395935
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395935 250 mg 250mg
30407828 100 mg 100mg
30394644 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395935 Supplier Toronto Research Chemicals Supplier No. TRCH951405250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material P-[2-Hydroxy-1-(phenylmethoxy)propyl]phosphonic Acid Dimethyl Ester
Analyte or Component Names P-[2-Hydroxy-1-(phenylmethoxy)propyl]phosphonic Acid Dimethyl Ester
Shipping Condition Room Temperature
Molecular Formula C12 H19 O5 P
InChI Formula InChI=1S/C12H19O5P/c1-10(13)12(18(14,15-2)16-3)17-9-11-7-5-4-6-8-11/h4-8,10,12-13H,9H2,1-3H3
Solution Type Neat
SMILES COP(=O)(OC)C(OCc1ccccc1)C(C)O
IUPAC Name 1-dimethoxyphosphoryl-1-phenylmethoxypropan-2-ol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 274.25
Quantity 250 mg
Formula Weight 274.097
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.