Learn More
Ozoralizumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA541947
Additional Details : Weight : 0.01000kg
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Ozoralizumab has been used in trials studying the treatment of Rheumatoid Arthritis and Active Rheumatoid Arthritis.Specifications
Ozoralizumab | |
Recombinant Monoclonal | |
Unconjugated | |
ATN-103 | |
Human ALB/TNFSF2/TNF-alpha/TNFA | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA | |
0.52 mg/mL | |
PBS with no preservative; pH 7.5 | |
Human | |
Metal-chelate chromatography | |
RUO | |
Chemical | |
Antibody | |
Chemical |