missing translation for 'onlineSavingsMsg'
Learn More

Ozenoxacin, MedChemExpress

Product Code. 16433456
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16433456

Brand: Medchem Express HY14957

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ozenoxacin is a nonfluorinated quinolone antibacterial, which shows potent activities against the main microorganisms isolated from skin and soft tissue infections.

Ozenoxacin is a nonfluorinated quinolone antibacterial, which shows potent activities against the main microorganisms isolated from skin and soft tissue infections.

Specifications

Chemical Name or Material Ozenoxacin
Physical Form Powder
CAS 245765-41-7
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C21H21N3O3
For Use With (Application) COVID-19-immunoregulation
Synonym T-3912
Solubility Information DMSO : 4.17 mg/mL (11.47 mM; ultrasonic and warming and heat to 70°C)
SMILES O=C(C1=CN(C2CC2)C3=C(C=CC(C4=CC(C)=C(NC)N=C4)=C3C)C1=O)O
Molecular Weight (g/mol) 363.41
Formula Weight 363.41
Percent Purity 98.58%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.