missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Oxytocin Receptor Recombinant Rabbit Monoclonal Antibody (7X8B8)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537974
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, PC-3, Jukat, Mouse brain Immunogen sequence: LLLALSGNAC VLLALRTTRQ KHSRLFFFMK HLSIADLVVA VFQVLPQLLW DITFRFYGPD LLCRLVKYLQ VVGMFASTYL LLLMSLDRCL AICQPLRSLR R.
The oxytocin receptor (OXTR) belongs to the G-protein coupled receptor family. Its activity is mediated by G proteins which activate a phosphatidylinositol-calcium second messenger system. The receptor is made up of 389 amino acids and has a molecular weight of ~43,000 kDa. The oxytocin-oxytocin receptor system plays an important role as an inducer of uterine contractions during parturition and of milk ejection. Oxytocin receptors are expressed by the myoepithelial cells of the mammary gland, and in both the myometrium and endometrium or the uterus at the end of pregnancy. Additionally, oxytocin receptors are present in the central nervous system, mediating a variety of behaviors, including maternal behavior, bonding, social memory and recognition, sexual and aggressive behaviors, and stress and anxiety.
Specifications
| Oxytocin Receptor | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P30559, P70536, P97926 | |
| Oxtr | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 7X8B8 | |
| Unconjugated | |
| Oxtr | |
| Otr; OT-R; OTR1; oxcytocin receptor; OXTR; Oxt-r; Oxytocin receptor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18430, 25342, 5021 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction