Learn More
Invitrogen™ Oxytocin Receptor Recombinant Rabbit Monoclonal Antibody (24GB560)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557143
 View more versions of this product
Product Code. 30275614
  • £434.00 / 100µL
Description
Oxytocin Receptor Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The oxytocin receptor (OXTR) belongs to the G-protein coupled receptor family. Its activity is mediated by G proteins which activate a phosphatidylinositol-calcium second messenger system. The receptor is made up of 389 amino acids and has a molecular weight of ~43,000 kDa. The oxytocin-oxytocin receptor system plays an important role as an inducer of uterine contractions during parturition and of milk ejection. Oxytocin receptors are expressed by the myoepithelial cells of the mammary gland, and in both the myometrium and endometrium or the uterus at the end of pregnancy. Additionally, oxytocin receptors are present in the central nervous system, mediating a variety of behaviors, including maternal behavior, bonding, social memory and recognition, sexual and aggressive behaviors, and stress and anxiety.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Oxytocin Receptor
Recombinant Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P30559, P70536, P97926
Oxtr
A synthesized peptide derived from human Oxytocin Receptor (aa 95-120).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB560
Unconjugated
Oxtr
Otr; OT-R; OTR1; oxcytocin receptor; OXTR; Oxt-r; Oxytocin receptor
Rabbit
Affinity chromatography
RUO
18430, 25342, 5021
-20°C