missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Oxytocin Receptor Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142214
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
The oxytocin receptor (OXTR) belongs to the G-protein coupled receptor family. Its activity is mediated by G proteins which activate a phosphatidylinositol-calcium second messenger system. The receptor is made up of 389 amino acids and has a molecular weight of ~43,000 kDa. The oxytocin-oxytocin receptor system plays an important role as an inducer of uterine contractions during parturition and of milk ejection. Oxytocin receptors are expressed by the myoepithelial cells of the mammary gland, and in both the myometrium and endometrium or the uterus at the end of pregnancy. Additionally, oxytocin receptors are present in the central nervous system, mediating a variety of behaviors, including maternal behavior, bonding, social memory and recognition, sexual and aggressive behaviors, and stress and anxiety.
Specifications
| Oxytocin Receptor | |
| Polyclonal | |
| Unconjugated | |
| Oxtr | |
| Otr; OT-R; OTR1; oxcytocin receptor; OXTR; Oxt-r; Oxytocin receptor | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 5021 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P30559 | |
| Oxtr | |
| Peptide with sequence C-LRTTRQKHSRL. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction