missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Oxytocin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575431
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
There are two proteins encoded by this gene, oxytocin and neurophysin I. Oxytocin is posterior pituitary hormone which is synthesized as an inactive precursor in the hypothalamus along with its carrier protein neurophysin I. Together with neurophysin, it is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis, where it is either stored or secreted into the bloodstream. The precursor seems to be activated while it is being transported along the axon to the posterior pituitary. This hormone contracts smooth muscle during parturition and lactation. It is also involved in cognition, tolerance, adaptation and complex sexual and maternal behaviour, as well as in the regulation of water excretion and cardiovascular functions.
Specifications
| Oxytocin | |
| Polyclonal | |
| Unconjugated | |
| OXT | |
| neurophysin 1; neurophysin I; Ocytocin; Ot; OT-NPI; oxcytocin; OXT; OXT-NPI; Oxy; Oxytocin; oxytocin, prepro- (neurophysin I); oxytocin, prepropeptide; oxytocin/neurophysin (Oxt) gene, complete gene, complete cds; oxytocin/neurophysin 1 prepropeptide; oxytocin/neurophysin I prepropeptide; oxytocin-neurophysin 1; oxytocin-neurophysin I; oxytocin-neurophysin I, preproprotein; prepropeptide for oxytocin and neurophysin; unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18429, 25504, 5020 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P01178, P01179, P35454 | |
| OXT | |
| Synthetic peptide corresponding to amino acids 11-60 of Human Oxytocin. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction