missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxytocin/Neurophysin I Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-76346
This item is not returnable.
View return policy
Description
Oxytocin/Neurophysin I Polyclonal specifically detects Oxytocin/Neurophysin I in Mouse, Rat samples. It is validated for Flow Cytometry, Peptide ELISA, Immunofluorescence.
Specifications
| Oxytocin | |
| Polyclonal | |
| Flow Cytometry, Peptide ELISA Detection limit 1:16000, Immunofluorescence | |
| MGC126890, MGC126892, neurophysin I, OT, OT-NPI, oxytocin, prepro- (neurophysin I), oxytocin, prepropeptide, oxytocin-neurophysin 1, oxytocin-neurophysin I, preproprotein | |
| Goat | |
| Affinity Purified | |
| Primary | |
| Mouse, Rat | |
| IgG |
| Flow Cytometry, ELISA, Immunofluorescence | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| OXT | |
| Peptide with sequence DGCRTDPACDPES corresponding to C-Terminus (near) according to Uniprot Mouse NP_035155.1. | |
| 0.1 mg | |
| 5020.0 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction