missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxytocin/Neurophysin I Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94626-0.1ml
This item is not returnable.
View return policy
Description
Oxytocin/Neurophysin I Polyclonal antibody specifically detects Oxytocin/Neurophysin I in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Oxytocin/Neurophysin I | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000 | |
| neurophysin I, OT, OT-NPI, OXT, OXT-NPI, oxytocin, prepro- (neurophysin I), oxytocin, prepropeptide, oxytocin-neurophysin 1, oxytocin-neurophysin I, preproprotein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human OXT (NP_000906.1). MAGPSLACCLLGLLALTSACYIQNCPLGGKRAAPDLDVRKCLPCGPGGKGRCFGPNICCAEELGCFVGTAEALRCQEENYLPSPCQSGQKACGSGGRCAVLGLCCSPDGCHADPACDAEATFSQR | |
| 0.1 mL | |
| Neuroscience | |
| 5020 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction