missing translation for 'onlineSavingsMsg'
Learn More

Oxytocin Acetate, TRC

Product Code. 30041366
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041366 100 mg 100mg
30041003 10 mg 10mg
30041317 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041366 Supplier Toronto Research Chemicals Supplier No. TRCO878500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6233-83-6

Specifications

Purity >95% (HPLC)
Format Neat
Molecular Formula C45H70N12O14S2
Molecular Weight (g/mol) 1067.24, 1067.25
InChI Key DSZOEVVLZMNAEH-BXUJZNQYSA-N
SMILES CC(O)=O.[H][C@]1(NC(=O)[C@H](CC2=CC=C(O)C=C2)NC(=O)[C@@H](N)CSSC[C@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CCC(N)=O)NC1=O)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CC(C)C)C(=O)NCC(N)=O)[C@@H](C)CC
IUPAC Name (2S)-2-{[(2S)-1-[(4R,7S,10S,13S,16S,19R)-19-amino-13-[(2S)-butan-2-yl]-10-(2-carbamoylethyl)-7-(carbamoylmethyl)-16-[(4-hydroxyphenyl)methyl]-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentaazacycloicosane-4-carbonyl]pyrrolidin-2-yl]formamido}-N-(carbamoylmethyl)-4-methylpentanamide; acetic acid
Product Type API
Quantity 100 mg
Shipping Condition Room Temperature
Synonym Oxytocin, acetate (1:1)∣Oxytocin, acetate (7CI)∣Oxytocin, monoacetate (salt) (8CI,9CI)∣1,2-Dithia-5,8,11,14,17-pentaazacycloeicosane, cyclic peptide deriv.∣Oxytocin acetate (salt)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.