missing translation for 'onlineSavingsMsg'
Learn More

Oxytetracycline Hydrochloride, LoGiCal

Product Code. 30459363
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459363 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459363 Supplier LoGiCal Supplier No. LGCFOR0554.00
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxytetracycline Hydrochloride
CAS 2058-46-0
Analyte or Component Names Oxytetracycline Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C22 H24 N2 O9 . Cl H
Certifications/Compliance ISO 9001
Synonym Oxytetracycline Hydrochloride, (4S,4aR,5S,5aR,6S,12aS)-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, 4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, [4S-(4alpha,4aalpha,5alpha,5aalpha,6beta,12aalpha)]-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride, 1-Dimethylamino-1,4,6,11,12,13,14,18-octahydro-2,5,7,11,12,14-hexahydroxy-4,6-dioxo-11-methylnaphthacene-3-carbonamide hydrochloride, 5-Hydroxytetracycline hydrochloride, 5-Hydroxytetracycline monohydrochloride, Alamycin, Aquacycline, Biocyl, Duphacycline, Engemycin, Gynamousse, Liquamycin Injectable, Maxicyklin, Mepatar, Occrycetin, Oxlopar, Oxy WS, Oxy-Dumocyclin, Oxybiocycline, Oxybiotic, Oxycyclin, Oxyject, Oxylag, Oxypan, Oxytet, Oxytetracid, Oxytetracycline hydrochloride, Oxytetrin, TM 5, Terraject, Terramycin hydrochloride, Tetra-Tabilinen, Tetramel, Tetran hydrochloride, Toxinal, Doxycycline Hyclate Impurity E as Hydrochloride, Doxycycline Impurity E as Hydrochloride
InChI Formula InChI=1S/C22H24N2O9.ClH/c1-21(32)7-5-4-6-8(25)9(7)15(26)10-12(21)17(28)13-14(24(2)3)16(27)11(20(23)31)19(30)22(13,33)18(10)29;/h4-6,12-14,17,25,27-29,32-33H,1-3H3,(H2,23,31);1H/t12-,13-,14+,17+,21-,22+;/m1./s1
Solution Type Neat
SMILES Cl.CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)cccc4[C@@]3(C)O
IUPAC Name (4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 496.89
Quantity 10 mg
Formula Weight 496.1249 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.