missing translation for 'onlineSavingsMsg'
Learn More

Oxytetracycline Dihydrate, TRC

Product Code. 30411141
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411141 10 g -
30411252 1 g -
30419140 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30411141 Supplier Toronto Research Chemicals Supplier No. TRCO87750110G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-64-6
Molecular Formula C22 H24 N2 O9 . 2 H2 O
Molecular Weight (g/mol) 496.46
Synonym Oxytetracycline dihydrate, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-, hydrate (1:2), (4S,4aR,5S,5aR,6S,12aS)-, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-, dihydrate (8CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-, dihydrate, (4S,4aR,5S,5aR,6S,12aS)- (9CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-, dihydrate, [4S-(4α,4aα,5α,5aα,6β,12aα)]-, (4S,4aR,5S,5aR,6S,12aS)-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide dihydrate, 5-Hydroxytetracycline dihydrate, Nitox
IUPAC Name (4S,4aR,5S,5aR,6S,12aS)-4-(dimethylamino)-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;dihydrate
SMILES O.O.CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)cccc4[C@@]3(C)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C22H24N2O9.2H2O/c1-21(32)7-5-4-6-8(25)9(7)15(26)10-12(21)17(28)13-14(24(2)3)16(27)11(20(23)31)19(30)22(13,33)18(10)29;;/h4-6,12-14,17,25,27-29,32-33H,1-3H3,(H2,23,31);2*1H2/t12-,13-,14+,17+,21-,22+;;/m1../s1
Formula Weight 496.1693
Chemical Name or Material Oxytetracycline dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.