missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oxytetracycline Dihydrate 95%

Product Code. 10631925 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10631925 10 g Glass Bottle 10g
10356910 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10631925 Supplier Thermo Scientific Chemicals Supplier No. 123840100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-64-6
Molecular Formula C22H28N2O11
Molecular Weight (g/mol) 496.469
MDL Number MFCD00151234
InChI Key IMLJLCJZQLGHJS-JEKSYDDFSA-N
PubChem CID 54680691
IUPAC Name (4S,4aR,5S,5aR,6S,12aR)-4-(dimethylamino)-1,5,6,10,11,12a-hexahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;dihydrate
SMILES CC1(C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O)O.O.O

Specifications

Type Oxytetracycline Dihydrate
Physical Form Crystalline Powder
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation On anhydrous substance: -203° to -216° (20°C, 589nm) (c=1, 0.1M HCl)
Water 4 to 8%
Color Yellow
Quantity 10 g
Assay Percent Range 95%
Merck Index 14, 6976
Formula Weight 496.46
Percent Purity ≥94% (on anhydrous substance)
Chemical Name or Material Oxytetracycline dihydrate, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Oxytetracycline dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.