missing translation for 'onlineSavingsMsg'
Learn More

Oxyquinoline (8-Hydroxyquinoline), Mikromol™

Product Code. 18063340
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18063340 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18063340 Supplier Mikromol Supplier No. MM3108.00
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Oxyquinoline (8-Hydroxyquinoline)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Hydroxyquinoline
CAS 148-24-3
Analyte or Component Names 8-Hydroxyquinoline
Shipping Condition Room Temperature
Molecular Formula C9 H7 N O
Certifications/Compliance ISO 17025
Synonym 8-Quinolinol, 1-Azanaphthalene-8-ol, 8-Hydroxychinolin, 8-Hydroxyquinoline, 8-OQ, 8-Oxyquinoline, 8-Quinol, AQ+, Albisal, Fennosan H 30, NSC 2039, NSC 285166, NSC 402623, NSC 48037, NSC 54230, NSC 615011, NSC 82404, NSC 82405, NSC 82409, NSC 82410, NSC 82412, Oxin, Oxine, Oxoquinoline, Oxychinolin, Oxyquinoline, Phenopyridine, Quinophenol, Tumex
InChI Formula InChI=1S/C9H7NO/c11-8-5-1-3-7-4-2-6-10-9(7)8/h1-6,11H
SMILES Oc1cccc2cccnc12
IUPAC Name quinolin-8-ol
Molecular Weight (g/mol) 145.16
Quantity 250 mg
Formula Weight 145.0528 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.