missing translation for 'onlineSavingsMsg'
Learn More

Oxyphenisatin acetate, MedChemExpress

Product Code. 16424666
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16424666 100 mg 100mg
16419966 50 mg 50mg
16403528 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16424666 Supplier Medchem Express Supplier No. HY101714
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oxyphenisatin acetate, the pro-drug of oxyphenisatin, is used to be a laxative.

Oxyphenisatin acetate, the pro-drug of oxyphenisatin, is used to be a laxative.

Specifications

Chemical Name or Material Oxyphenisatin acetate
CAS 115-33-3
Color White
Purity Grade Notes Research
Molecular Formula C24H19NO5
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (249.12 mM; Need ultrasonic)
SMILES O=C1NC2=C(C=CC=C2)C1(C3=CC=C(OC(C)=O)C=C3)C4=CC=C(OC(C)=O)C=C4
Molecular Weight (g/mol) 401.41
Formula Weight 401.41
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.