missing translation for 'onlineSavingsMsg'
Learn More

Oxypaeoniflorin, MedChemExpress

Product Code. 16431498
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16431498

Brand: Medchem Express HYN0748

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oxypaeoniflorin, an anti-oxidant, is a monoterpene glycoside compound isolated from Paeoniae species. Oxypaeoniflorin has neuroprotective and anti-inflammatory effects.

Oxypaeoniflorin, an anti-oxidant, is a monoterpene glycoside compound isolated from Paeoniae species. Oxypaeoniflorin has neuroprotective and anti-inflammatory effects.

Specifications

Chemical Name or Material Oxypaeoniflorin
CAS 39011-91-1
Color White
Purity Grade Notes Research
Molecular Formula C23H28O12
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (201.43 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(O)C=C1)OC[C@]([C@@]([C@]2(O)O3)([H])C4)([C@]3([H])O[C@]5(C2)C)[C@]45O[C@]([C@@H]([C@@H](O)[C@@H]6O)O)([H])O[C@@H]6CO
Molecular Weight (g/mol) 496.46
Formula Weight 496.46
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.