missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Oxymatrine, Dr. Ehrenstorfer
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Oxymatrine |
| CAS | 16837-52-8 |
| Analyte or Component Names | Oxymatrine |
| Shipping Condition | +20°C |
| Molecular Formula | C15 H24 N2 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 1H,5H,10H-Dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one, dodecahydro-, 4-oxide, (4R,7aS,13aR,13bR,13cS)-, Matridin-15-one, 1-oxide, (1β)-, Matrine, 1-oxide (8CI), Matrine, N-oxide (6CI,7CI), (+)-Matrine N-oxide, (+)-Oxymatrine, 1H,5H,10H-Dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one, dodecahydro-, 4-oxide, [4R-(4α,7aβ,13aα,13bβ,13cβ)]-, Ammothamnine, Matrine 1β-oxide, Matrine N1-oxide, Oxymatrine |
| InChI Formula | InChI=1S/C15H24N2O2/c18-14-7-1-6-13-12-5-3-9-17(19)8-2-4-11(15(12)17)10-16(13)14/h11-13,15H,1-10H2/t11-,12+,13+,15-,17?/m0/s1 |
| SMILES | [O-][N+]12CCC[C@H]3CN4[C@H](CCCC4=O)[C@@H](CCC1)[C@@H]23 |
| Molecular Weight (g/mol) | 264.36 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction