missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Oxymatrine

Product Code. 16431496
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16431496 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16431496 Supplier MedChemExpress Supplier No. HYN0158
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oxymatrine, an alkaloid from the roots of Sophora species, with anti-inflammatory, antifibrosis, and antitumor effects, inhibits the iNOS expression and TGF-β/Smad pathway. Oxymatrine inhibits bocavirus minute virus of canines (MVC) replication, reduces viral gene expression and decreases apoptosis induced by viral infection.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 264.36 g/mol
Shipping Condition Room temperature
For Use With (Application) COVID-19-anti-virus
CAS 16837-52-8
Product Type Natural Products
Purity 98.0%
Solubility H2O : 100 mg/mL (378.27 mM∣Need ultrasonic) ∣DMSO : 100 mg/mL (378.27 mM∣Need ultrasonic)
Target TGF-beta/Smad∣Influenza Virus
Quantity 200 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.