missing translation for 'onlineSavingsMsg'
Learn More

Oxyclozanide, MedChemExpress

Product Code. 16457386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16457386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16457386 Supplier Medchem Express Supplier No. HY17594
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oxyclozanide is a salicylanilide anthelmintic drug that mainly acts by uncoupling oxidative phosphorylation in flukes.

Oxyclozanide is a salicylanilide anthelmintic drug that mainly acts by uncoupling oxidative phosphorylation in flukes.

Specifications

Chemical Name or Material Oxyclozanide
CAS 2277-92-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C13H6Cl5NO3
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (249.09 mM; Need ultrasonic)
SMILES O=C(NC1=CC(Cl)=CC(Cl)=C1O)C2=C(O)C(Cl)=CC(Cl)=C2Cl
Molecular Weight (g/mol) 401.46
Formula Weight 401.46
Percent Purity 98.85%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.