missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxybutynin EP Impurity D, Mikromol™
Description
Oxybutynin EP Impurity D
Specifications
Specifications
| Chemical Name or Material | Phenylcyclohexylglycolic Acid |
| CAS | 4335-77-7 |
| Analyte or Component Names | Phenylcyclohexylglycolic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H18 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetic Acid (Phenylcyclohexylglycolic Acid), Oxybutynin Hydrochloride Imp. D (EP), Oxybutynin Imp. D (EP), Oxybutynin USP Related Compound A, Oxybutynin USP RC A, (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetic Acid, Phenylcyclohexylglycolic Acid, Oxybutynin Related Compound A, Oxybutynin Hydrochloride Impurity D, Oxybutynin Impurity D |
| InChI Formula | InChI=1S/C14H18O3/c15-13(16)14(17,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1,3-4,7-8,12,17H,2,5-6,9-10H2,(H,15,16) |
| SMILES | OC(=O)C(O)(C1CCCCC1)c2ccccc2 |
| IUPAC Name | 2-cyclohexyl-2-hydroxy-2-phenylacetic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?