missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
OXTR Polyclonal antibody specifically detects OXTR in Human, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | OXTR |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA, Immunohistochemistry 40 μg/mL, Immunohistochemistry-Paraffin 40 μg/mL |
| Formulation | PBS |
| Gene Accession No. | P30559 |
| Gene Alias | OT-R, oxytocin receptor |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?