missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXSR1 Antibody (OTI1F3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47905
This item is not returnable.
View return policy
Description
OXSR1 Monoclonal specifically detects OXSR1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| OXSR1 | |
| Monoclonal | |
| 1.28 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
| OXSR1 | |
| Full-length protein expressed in 293T cell transfected with human OXSR1 expression vector | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific for Homo sapiens oxidative-stress responsive 1 (OXSR1). | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI1F3 | |
| Unconjugated | |
| EC 2.7.11, EC 2.7.11.1, KIAA1101OSR1serine/threonine-protein kinase OSR1, Oxidative stress-responsive 1 protein, oxidative-stress responsive 1 | |
| Mouse | |
| 57.8 kDa | |
| 0.1 mL | |
| Protein Kinase | |
| 9943 | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction