missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXSR1 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-77321IR
This item is not returnable.
View return policy
Description
OXSR1 Polyclonal antibody specifically detects OXSR1 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| OXSR1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.11, EC 2.7.11.1, KIAA1101OSR1serine/threonine-protein kinase OSR1, Oxidative stress-responsive 1 protein, oxidative-stress responsive 1 | |
| The immunogen recognized by this antibody maps to a region between residue 325 and 375 of human oxidative-stress responsive 1 using the numbering given in entry NP_005100.1 (GeneID 9943). | |
| 0.1 mL | |
| Protein Kinase | |
| 9943 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction