missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ OXPAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117687
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
This antibody has been tested in direct-ELISA.
Lipid droplet-associated protein that maintains the balance between lipogenesis and lipolysis and also regulates fatty acid oxidation in oxidative tissues. Recruits mitochondria to the surface of lipid droplets and is involved in lipid droplet homeostasis by regulating both the storage of fatty acids in the form of triglycerides and the release of fatty acids for mitochondrial fatty acid oxidation. In lipid droplet triacylglycerol hydrolysis, plays a role as a scaffolding protein for three major key lipolytic players: ABHD5, PNPLA2 and LIPE. Reduces the triacylglycerol hydrolase activity of PNPLA2 by recruiting and sequestering PNPLA2 to lipid droplets. Phosphorylation by PKA enables lipolysis probably by promoting release of ABHD5 from the perilipin scaffold and by facilitating interaction of ABHD5 with PNPLA2. Also increases lipolysis through interaction with LIPE and upon PKA-mediated phosphorylation of LIPE.
Tekniske data
| OXPAT | |
| Polyclonal | |
| Unconjugated | |
| Plin5 | |
| 2310076L09Rik; AI415325; AW109675; lipid droplet associated protein; lipid droplet-associated protein PAT-1; Lipid storage droplet protein 5; LSDA5; LSDP5; Mldp; Myocardial LD protein; Oxpat; Pat1; PAT-1; perilipin 5; perilipin-5; Plin5 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein G | |
| RUO | |
| 440503, 66968 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300, 0.02% sodium azide; pH 7.4 | |
| Q00G26, Q8BVZ1 | |
| Plin5 | |
| Recombinant Human Perilipin-5 protein (305-453aa). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion