missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OXPAT Polyclonal Antibody
Description
The target sequence has 89% sequence homology with rat and 75% sequence homology with bovine. Suggested positive control: human and mouse heart lysate.
Lipid droplet-associated protein that maintains the balance between lipogenesis and lipolysis and also regulates fatty acid oxidation in oxidative tissues. Recruits mitochondria to the surface of lipid droplets and is involved in lipid droplet homeostasis by regulating both the storage of fatty acids in the form of triglycerides and the release of fatty acids for mitochondrial fatty acid oxidation. In lipid droplet triacylglycerol hydrolysis, plays a role as a scaffolding protein for three major key lipolytic players: ABHD5, PNPLA2 and LIPE. Reduces the triacylglycerol hydrolase activity of PNPLA2 by recruiting and sequestering PNPLA2 to lipid droplets. Phosphorylation by PKA enables lipolysis probably by promoting release of ABHD5 from the perilipin scaffold and by facilitating interaction of ABHD5 with PNPLA2. Also increases lipolysis through interaction with LIPE and upon PKA-mediated phosphorylation of LIPE.
Specifications
Specifications
| Antigen | OXPAT |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.01 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.1% sodium azide |
| Gene | Plin5 |
| Gene Accession No. | Q00G26, Q8BVZ1 |
| Gene Alias | 2310076L09Rik; AI415325; AW109675; lipid droplet associated protein; lipid droplet-associated protein PAT-1; Lipid storage droplet protein 5; LSDA5; LSDP5; Mldp; Myocardial LD protein; Oxpat; Pat1; PAT-1; perilipin 5; perilipin-5; Plin5 |
| Gene Symbols | Plin5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?