missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXPAT Antibody, Allophycocyanin, Novus Biologicals™
Description
OXPAT Polyclonal antibody specifically detects OXPAT in Human, Mouse, Porcine, Bovine samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
Specifications
| Applications | Western Blot |
| Target Species | Pig |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?