missing translation for 'onlineSavingsMsg'
Learn More

Oxophenylacetic Acid (Benzoylformic Acid), Mikromol™

Product Code. 30051636
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051636 25 mg 25mg
30049545 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051636 Supplier Mikromol Supplier No. MM0834.050025
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Oxophenylacetic Acid (Benzoylformic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenylglyoxylic Acid
CAS 611-73-4
Analyte or Component Names Phenylglyoxylic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H6 O3
Certifications/Compliance ISO 9001
Synonym Glycopyrronium Bromide Imp. B (EP), Oxophenylacetic Acid, Benzoylformic Acid, Glycopyrronium Bromide Impurity B, Glycopyrronium Impurity B
InChI Formula InChI=1S/C8H6O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5H,(H,10,11)
SMILES OC(=O)C(=O)c1ccccc1
IUPAC Name 2-oxo-2-phenylacetic acid
Molecular Weight (g/mol) 150.131
Quantity 25 mg
Formula Weight 150.032 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.