missing translation for 'onlineSavingsMsg'
Learn More

Oxonic Acid Potassium Salt, TRC

Product Code. 30409901
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409901 5 g 5g
30420124 25 g -
30423135 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30409901 Supplier Toronto Research Chemicals Supplier No. TRCO8700605G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2207-75-2
Molecular Formula C4 H2 N3 O4 . K
Molecular Weight (g/mol) 195.17
IUPAC Name potassium;4,6-dioxo-1H-1,3,5-triazine-2-carboxylate
SMILES [K+].[O-]C(=O)C1=NC(=O)NC(=O)N1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H3N3O4.K/c8-2(9)1-5-3(10)7-4(11)6-1;/h(H,8,9)(H2,5,6,7,10,11);/q;+1/p-1
Formula Weight 194.9682
Chemical Name or Material Oxonic Acid Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.