missing translation for 'onlineSavingsMsg'
Learn More

Oxonic acid potassium salt

Product Code. 15493169
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493169 25 g 25g
15483169 5 g 5g
15403179 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15493169 Supplier Thermo Scientific Alfa Aesar Supplier No. J60400.14
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of uricase | CAS: 2207-75-2 | C4H2KN3O4 | 195.18 g/mol

Oxonic acid potassium salt is an inhibitor of uricase. This product has been used for the inhibition of 5-fluorouracil-induced gastrointestinal toxicity without the loss of its antitumor activity in rats. It has also been used to induce hyperuricemia in rats; as it inhibits uric acid metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxonic acid potassium salt is an inhibitor of uricase. This product has been used for the inhibition of 5-fluorouracil-induced gastrointestinal toxicity without the loss of its antitumor activity in rats. It has also been used to induce hyperuricemia in rats; as it inhibits uric acid metabolism.

Solubility
Soluble in hot water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2207-75-2
Molecular Formula C4H2KN3O4
Molecular Weight (g/mol) 195.18
MDL Number MFCD00010565
InChI Key IAPCTXZQXAVYNG-UHFFFAOYSA-M
Synonym potassium oxonate, oteracil potassium, oxonic acid potassium salt, allantoxanic acid potassium salt, potassium azaorotate, oxonic acid, potassium salt, oxonate, potassium, oxonate, allantoxanic acid
PubChem CID 2723920
ChEBI CHEBI:80230
SMILES [K+].[O-]C(=O)C1=NC(=O)NC(=O)N1

Specifications

Melting Point 300°C
Color White
Physical Form Powder
Quantity 25 g
Merck Index 14,6947
Solubility Information Soluble in hot water.
IUPAC Name potassium 4,6-dioxo-1,4,5,6-tetrahydro-1,3,5-triazine-2-carboxylate
Formula Weight 195.18
Chemical Name or Material Oxonic acid potassium salt, 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.