missing translation for 'onlineSavingsMsg'
Learn More

Oxolinic Acid, TRC

Product Code. 30411838
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411838 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411838 Supplier Toronto Research Chemicals Supplier No. TRCO8575005G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxolinic acid
CAS 14698-29-4
Molecular Formula C13 H11 N O5
Quantity 5 g
Synonym Oxolinic Acid, 1-Ethyl-1,4-dihydro-6,7-methylenedioxy-4-oxo-3-quinolinecarboxylic acid, 1-Ethyl-6,7-methylenedioxy-4-quinolone-3-carboxylic acid, 5-Ethyl-5,8-dihydro-8-oxo-1,3-dioxolo[4,5-g]quinoline-7-carboxylic acid, Dioxacin, Emyrenil, Gramurin, Inoxyl, NSC 110364, Nidantin, Ossian, Oxoboi, Oxolinic acid, Pietil, Prodoxal, Prodoxol, S 0208, Starner, Starner 20WP, Ultibid, Urinox, Uritrate, Uro-Alvar, Urotrate, Uroxin, Uroxol, Utibid, W 4565
InChI Formula InChI=1S/C13H11NO5/c1-2-14-5-8(13(16)17)12(15)7-3-10-11(4-9(7)14)19-6-18-10/h3-5H,2,6H2,1H3,(H,16,17)
SMILES CCN1C=C(C(=O)O)C(=O)c2cc3OCOc3cc12
IUPAC Name 5-ethyl-8-oxo-[1,3]dioxolo[4,5-g]quinoline-7-carboxylic acid
Molecular Weight (g/mol) 261.23
Formula Weight 261.0637
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.