missing translation for 'onlineSavingsMsg'
Learn More

Oxolamine Citrate, TRC

Product Code. 30012392
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012392 25 g 25g
30005987 10 g 10g
30004035 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012392 Supplier Toronto Research Chemicals Supplier No. TRCO85745025G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1949-20-8
Molecular Formula C14 H19 N3 O . C6 H8 O7
Molecular Weight (g/mol) 437.44
Synonym 1,2,4-Oxadiazole-5-ethanamine, N,N-diethyl-3-phenyl-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), 1,2,4-Oxadiazole, 5-[2-(diethylamino)ethyl]-3-phenyl-, citrate (1:1) (7CI,8CI), 3-Phenyl-5-(diethylaminoethyl)-1,2,4-oxadiazole citrate, 5-(Diethylaminoethyl)-3-phenyl-1,2,4-oxadiazole citrate, AF 438, Bredon, Broncatar, Flogobron, NSC 100298, Oxarmin, Oxolamine citrate, Perebron, Prilon, SKF 9976
IUPAC Name N,N-diethyl-2-(3-phenyl-1,2,4-oxadiazol-5-yl)ethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCN(CC)CCc1onc(n1)c2ccccc2.OC(=O)CC(O)(CC(=O)O)C(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C14H19N3O.C6H8O7/c1-3-17(4-2)11-10-13-15-14(16-18-13)12-8-6-5-7-9-12;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-9H,3-4,10-11H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)
Formula Weight 437.1798
Chemical Name or Material Oxolamine Citrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.