missing translation for 'onlineSavingsMsg'
Learn More

Oxolamine citrate, MedChemExpress

Product Code. 16412309
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16412309 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16412309 Supplier Medchem Express Supplier No. HYB1042
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oxolamine citrate (SKF-9976 citrate) is a cough suppressant that can be used for the research of respiratory tract diseases. Oxolamine citrate also exhibits anti-inflammatory effect.

Oxolamine citrate (SKF-9976 citrate) is a cough suppressant that can be used for the research of respiratory tract diseases. Oxolamine citrate also exhibits anti-inflammatory effect.

Specifications

Chemical Name or Material Oxolamine citrate
Physical Form Solid
CAS 1949-20-8
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C20H27N3O8
For Use With (Application) COVID-19-immunoregulation
Synonym SKF-9976 citrate AF-438 citrate
Solubility Information DMSO : ≥ 100 mg/mL (228.60 mM) ∣H2O : 14.29 mg/mL (32.67 mM; Need ultrasonic)
SMILES CCN(CC)CCC1=NC(C2=CC=CC=C2)=NO1.O=C(CC(C(O)=O)(O)CC(O)=O)O
Molecular Weight (g/mol) 437.44
Formula Weight 437.44
Percent Purity 99.55%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.