Learn More
Thermo Scientific™ Oxoid™ Ceftazidime/Avibactam Antimicrobial Susceptibility discs
Product Code. 16391438
missing translation for 'orderingAttributeHoverText'
Concentration:
10/4 μg
missing translation for 'unitSize'
Pack of 250
Product Code. 16391438
missing translation for 'mfr': Thermo Scientific™ CT1948B
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality antimicrobial susceptibility testing discs.
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Ceftazidime/Avibactam Antimicrobial Susceptibility discs. Ceftazidime/Avibactam is indicated for the treatment of complicated urinary tract infections, including pyelonephritis, in patients 18 years or older. In combination with metronidazole, it is also indicated for the treatment of complicated intra-abdominal infections.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling, and end-of-cartridge indicators.
Disc diffusion not only categorizes resistant, intermediate, and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Oxoid Ceftazidime/Avibactam Antimicrobial Susceptibility discs are available in two concentrations to meet CLSI or EUCAST recommended levels. Oxoid discs are manufactured in accordance with appropriate quality standards, including BS EN ISO 9001:2008, ISO 13485:2003 and FDA 21 CFR part 820, and undergo comprehensive quality control testing to ensure maximum accuracy and reproducibility.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Ceftazidime/Avibactam
Concentration 10/4 μg
Description Oxoid Ceftazidime/Avibactam Antimicrobial Susceptibility Discs
Antibiotic Code CZA
Product Type Antimicrobial Susceptibility Discs
Test Type Antimicrobial Susceptibility
No. per Pack 5 Cartridges/Pk.
Packaging Type 50 Discs/Cartridge
Quantity 50 Discs/Cartridge
CE IVD status