missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Wort Agar (Dehydrated)

Product Code. 10556133
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10556133

Brand: Thermo Scientific™ CM0247B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cultivate and enumerate yeasts with this general purpose mycological medium.

Cultivate and enumerate yeasts with general purpose mycological Thermo Scientific™ Oxoid™ Wort Agar (Dehydrated). The medium equivalent to the medium described by Parfitt1 duplicates the acidity of natural wort making it optimal for many yeasts but inhibitory to most bacteria.

Wort Agar is a general purpose mycological medium, equivalent to the medium described by Parfitt1 and especially suitable for the cultivation and enumeration of yeasts. The medium, which duplicates the composition of natural wort, is of an acidity which is optimal for many yeasts but inhibitory to most bacteria. Parfitt investigated the relative merits of wort agar and other media for the count of yeasts and molds in butter, and recommended the use of dehydrated whey, malt or wort agar for the purpose. Scarr2 employed a modified wort agar (’osmophilic agar’) for the examination of sugar products for osmophilic yeasts. Scarr’s technique is also used for the determination of osmophilic yeasts occurring in materials used in the manufacture of soft drinks.

TRUSTED_SUSTAINABILITY

Specifications

Description Wort Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 10.0L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.