missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Wilkins-Chalgren Anaerobe Broth (Dehydrated)

Product Code. 10689744
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10689744 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10689744 Supplier Thermo Scientific™ Supplier No. CM0643B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate, cultivate and /or carry out susceptibility testing of a wide variety of anaerobic organisms from clinical samples with this medium.

Isolate, cultivate and/or carry out susceptibility testing of a wide variety of anaerobic organisms from clinical samples with Thermo Scientific™ Oxoid™ Wilkins-Chalgren Anaerobe Blood Agar (Dehydrated). Wilkins-Chalgren Anaerobe Broth supports the luxuriant growth of anaerobic organisms. It is derived from Wilkins Charlgren Agar1 CM0619. Where studies on antimicrobial susceptibilities are to be made in both broth and agar, this medium is found to be of particular value.

Use Anaerobe Blood Broth for the isolation of general anaerobes from clinical samples and for antimicrobial susceptibility testing of aerobic bacteria using Receiver Operating Characteristic (ROC) procedure.

The formulation of Wilkins-Chalgren Anaerobe Broth includes yeast extract to supply vitamins and other growth factors such as purines and pyrimidines that are necessary for good growth of Peptostreptococcus anaerobius and Bacteroides melaninogenicus. Arginine is added to ensure sufficient amino-acid is available for the growth of Eubacterium lentum. Pyruvate is present as an energy source for asaccharolytic cocci such as Veillonella. It also acts similarly to catalase and degrades traces of hydrogen peroxide, which may be produced by the action of molecular oxygen on medium constituents and interfere with the metabolism of anaerobes. Haemin is found to be essential for the growth of Bacteroides species and menadione for Bacteroides melaninogenicus.

TRUSTED_SUSTAINABILITY

Specifications

Description Wilkins-Chalgren Anaerobe Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 15.2L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.