missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Vegetable Peptone Phosphate Broth
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
Thermo Scientific™ Oxoid Vegetable Peptone Phosphate Broth is a meat-free alternative to Tryptose Phosphate Broth.
- Suitable for use as a component of tissue culture media and for the cultivation of fastidious microorganisms
Specifications
Specifications
| Description | Vegetable Peptone Phosphate Broth |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 16.9L medium |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction