Learn More
Thermo Scientific™ Oxoid™ VCN Selective Supplement
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Description
Oxoid VCN Selective Supplement is used for the preparation of Thayer Martin Medium for the isolation of pathogenic Neisseria spp.
- Add to GC Agar Base, Part No. CM0367B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | VCN Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |