Learn More
Thermo Scientific™ Oxoid™ Temocillin Antimicrobial Susceptibility Disks
Product Code. 15681344
Change view
Click to view available options
Concentration:
30 μg
Unit Size:
Pack of 5
Product Code. Concentration Unit Size
15681344 30 μg Pack of 5
1 options
Product Code. 15681344 Supplier Thermo Scientific™ Supplier No. CT1952B
  • ONLINE EXCLUSIVE PRICE
    £25.20
    £22.93 / Pack of 5
    Save £2.27
    9% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing disks.
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Temocillin Antimicrobial Susceptibility Disks.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
Disk diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Temocillin
Certifications/Compliance Complies with internally set guidelines to CLSI methodology.
Concentration 30 μg
Description Oxoid Temocillin Antimicrobial Susceptibility Disks
Antibiotic Code TEM
Product Type Antimicrobial Susceptibility Discs
Test Type Antimicrobial Susceptibility
No. per Pack 5 Cartridges/Pk.
Packaging Type 50 Disks/Cartridge
Quantity 50 Discs/Cartridge
For In Vitro Diagnostic Use.