missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ SS Agar Modified

Product Code. 10528643
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10528643 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10528643 Supplier Thermo Scientific™ Supplier No. CM0533B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selectively isolate, differentiate Shigella and Salmonella species from pathological specimens and suspected foodstuffs with this medium.

Selectively isolate, differentiate Shigella and Salmonella species from pathological specimens and suspected foodstuffs with Thermo Scientific™ Oxoid™ SS Agar Modified (Dehydrated). SS Agar Modified is designed to overcome the difficulties faced by SS agar. The improved formulation includes an alteration to the bile salt mixture, peptone, and pH value, improving its performance in the growth of shigellae and improved colony size for salmonellae with improved blackening at the center.

Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water. SS Agar Modified is ideal for screening samples containing mixed flora and suspected of harboring enteric pathogens, e.g., medical specimens or food products.

  • Easy-to-read: shigellae form transparent colonies while salmonellae form transparent colonies with black centers
  • Selective: selective inhibitory components brilliant green, bile salts, thiosulphate, and citrate inhibit Gram-positive organisms and non-target coliforms.

Although widely used, SS Agar has been criticized because of excessive inhibition of Shigella species.

Investigation has shown that modification to the formulation by alterations to the bile salt mixture, peptone and pH value considerably improve its performance in the growth of shigellae without too much increased growth of commensal organisms.

Salmonella colonies are also larger with improved blackening at the center. The change in formulation has reduced the number of g/L from 63g to 57g.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description SS Agar Modified
Form Powder
Product Type Agar
Quantity 500 g
Yield For 8.8L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.