missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Schaedler Anaerobe Broth (Dehydrated)

Product Code. 10679744
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10679744 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10679744 Supplier Thermo Scientific™ Supplier No. CM0497B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Grow and isolate aerobic and anaerobic organisms of the intestinal tract and/or carry out MIC studies with this thioglycollate free medium.

Grow and isolate aerobic and anaerobic organisms of the intestinal tract and/ or carry out minimum inhibitory concentration (MIC) studies of these organisms using Thermo Scientific™ Oxoid™ Schaedler Anaerobe Broth (Dehydrated).

Schandler Anaerobe Broth is free from thioglycollate and is used to create selective conditions under which delicate and more nutritionally exacting microorganisms of the intestinal tract develop, despite the presence of antagonistic organisms. Normally such fastidious microorganisms would be swamped by the growth of enterococci, coliform, bacilli, and other Gram-negative bacilli.

  • Selective against non-target Gram-negative bacteria, especially Escherichia coli
  • Can also be used to determine MIC levels of anaerobic organisms.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Schaedler Anaerobe Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 18.9L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.