missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Prepared Sabouraud Dextrose Broth Bottled Media

Product Code. 12766899
Change view
Click to view available options
Fill Weight:
10.0 to 10.3 g
100-101.5g
9.5 to 10.5 g
Format:
100 mL DIN bottle
Tube
Universal Bottle
Quantity:
10/Pk.
24/Pk.
50/Pk.
Unit Size:
Each
Pack of 24
Pack of 50
Product Code. Fill Weight Format Quantity unitSize
12766899 100-101.5g 100 mL DIN bottle 10/Pk. Each
10045972 10.0 to 10.3 g Universal Bottle 24/Pk. Pack of 24
13225809 9.5 to 10.5 g Tube 50/Pk. Pack of 50
3 options
This item is not returnable. View return policy

Product Code. 12766899

Brand: Thermo Scientific™ BO0283M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Confidently perform sterility testing and/or determining the fungistatic activity of pharmaceutical products with this broth.

A non-selective broth for confidently performing sterility testing and/or determining the fungistatic activity of pharmaceutical products.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2 to 25°C
Format 100 mL DIN bottle
Manufacturing Origin United Kingdom
Product Type Broth
Fill Weight 100-101.5g
Packaging Type 10 DIN bottles with wide septum, injectable closures in a correx box.
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.