missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Perfringens SFP Supplement

Product Code. 11464169
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464169 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11464169 Supplier Thermo Scientific™ Supplier No. SR0093E
  • ONLINE EXCLUSIVE PRICE
    £149.00
    £73.52 / Pack of 10
    Save £75.48
    50% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Oxoid Perfringens SFP Supplement is for the selective isolation of Clostridium perfringens.

Oxoid Perfringens SFP Supplement is for the selective isolation of Clostridium perfringens.

  • Add to Perfringens Agar Base, TSC, Part No. CM0587B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY

Specifications

Description Perfringens SFP Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Perfringens
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.