missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Perfringens OPSP Selective Supplement A
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Description
Oxoid Perfringens OPSP Selective Supplement A is for selective isolation of Clostridium perfringens from food.
- Add to Perfringens Agar Base, OPSP, Part No. CM0543B
- For 500mL of medium
Specifications
Specifications
| Description | Perfringens OPSP Supplement A |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Perfringens |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction