missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Perfringens agar Base (OPSP) (Dehydrated)

Product Code. 12771550
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.50kg
500g
This item is not returnable. View return policy

Product Code. 12771550

Brand: Thermo Scientific™ CM0543R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enumerate Clostridium perfringens in foods with this selective medium.

Enumerate Clostridium perfringens in foods with Thermo Scientific™ Oxoid™ Perfringens OPSP Agar Base (Dehydrated) when used with Perfringes (OPSP) Selective Supplements –A & B (SR0076 and SR0077). Perfringens OPSP Agar Base, is based on the formulation developed by Handford1.The medium utilizes sulphadiazine, oleandomycin phosphate and polymyxin B sulphate (10IU/mL), presented as freeze-dried supplements (SR0076 and SR0077) to give a high degree of selectivity and specificity for Clostridium perfringens.

Also available Perfringens OPSP Supplement A, Part No. SR0076E and Perfringens OPSP Supplement B, Part No. SR0077E.
TRUSTED_SUSTAINABILITY

Specifications

Description Perfringens Agar Base (OPSP)
Form Powder
Product Type Agar
Quantity 2.5 kg
Yield for 55.0L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.