Learn More
Thermo Scientific™ Oxoid™ Peptone P
Product Code. 10158822
Click to view available options
Quantity:
500g
Unit Size:
500g
Product Code. 10158822
Brand: Thermo Scientific™ LP0049B
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ Oxoid Peptone P is a peptic digest which conforms to the USP specification of peptic digests of animal tissue.
Thermo Scientific™ Oxoid Peptone P is a peptic digest which conforms to the USP specification of peptic digests of animal tissue.
  • The high sulphur content makes it ideal for use in culture media
TRUSTED_SUSTAINABILITY
Specifications
Description Dehydrated; Peptone P
Form Powder
Product Type Peptone
Quantity 500g