missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nutrient Gelatin (Dehydrated)
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
Differentiate microorganisms by their proteolytic effects with Thermo Scientific™ Oxoid™ Nutrient Gelatin (Dehydrated). Certain Enterobacteriaceae such as Proteus species and Serratia species have the ability to produce proteolytic enzyme that liquifies the gelatin. Therefore, nutrient gelatin is employed for differentiation of micro-organisms by their proteolytic effects.
Differentiate gelatin liquifying micro-organisms by 20°C plate count method with Nutrient Gelatin Agar.
Specifications
Specifications
| Description | Nutrient Gelatin (CM135a) |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Quantity | 500 g |
| Yield | For 3.9L medium |
For Laboratory Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction