Learn More
Thermo Scientific™ Oxoid™ Nutrient Agar Tablets
Product Code. 12897192
Change view
Click to view available options
Quantity:
Each
Unit Size:
100 tablets
Product Code. Quantity unitSize
12897192 Each 100 tablets
1 options
Product Code. 12897192 Supplier Thermo Scientific™ Supplier No. CM0004C
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Oxoid Nutrient Agar Tablets make 5mL of medium each.
Oxoid Nutrient Agar Tablets make 5mL of medium each.
TRUSTED_SUSTAINABILITY
Specifications
Description Nutrient Agar Tablets
Form Tablet
Product Type Nutrient Agar
Quantity Each
Yield Each tab for 5mL medium
For Laboratory Use Only